Tisk…

2-Cyclopropyl-1,3-thiazole-5-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (5-Carboxy-1,3-thiazol-2-yl)cyclopropane, 5-Carboxy-2-cyclopropyl-1,3-thiazole

OR41020-100MG OR41020-1G OR41020-250MG
APOSOR41020-100MGEA 4960 CZK
APOSOR41020-100MG APOSOR41020-1G APOSOR41020-250MG
2-Cyclopropyl-1,3-thiazole-5-carboxylic acid
2-Cyclopropyl-1,3-thiazole-5-carboxylic acid
Vzorec: C₇H₇NO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD11208434
CAS číslo: 1094230-05-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR