3-Chloro-4-(methylsulphonyl)thiophene-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4095-1G
OR4095-250MG
APOSOR4095-1GEA
6620
CZK
APOSOR4095-1G
APOSOR4095-250MG
3-Chloro-4-(methylsulphonyl)thiophene-2-carboxylic acid
3-Chloro-4-(methylsulphonyl)thiophene-2-carboxylic acid
Vzorec:
C₆H₅ClO₄S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00067947 CAS číslo: 175201-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...