(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR40822-1G
OR40822-250MG
OR40822-5G
APOSOR40822-1GEA
9100
CZK
APOSOR40822-1G
APOSOR40822-250MG
APOSOR40822-5G
(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester 95%
(4-Prop-2-ynoxyphenyl)boronic acid, pinacol ester
Vzorec:
C₁₅H₁₉BO₃ MW: 258,121 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD32177102 CAS číslo: 2280980-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...