Tisk…

1-Bromo-3-prop-2-ynoxy-benzene 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-3-(prop-2-yn-1-yloxy)benzene

OR40820-1G OR40820-250MG OR40820-5G
APOSOR40820-1GEA 992 CZK
APOSOR40820-1G APOSOR40820-250MG APOSOR40820-5G
1-Bromo-3-prop-2-ynoxy-benzene 97%
1-Bromo-3-prop-2-ynoxy-benzene
Vzorec: C₉H₇BrO
MW: 211,055 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09928819
CAS číslo: 114855-35-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR