Tisk…

2-Bromo-5-(cyclopropyl)thiophene

Dodavatel: Apollo Scientific
Varování

OR40806-1G OR40806-250MG OR40806-5G
APOSOR40806-1GEA 9920 CZK
APOSOR40806-1G APOSOR40806-250MG APOSOR40806-5G
2-Bromo-5-(cyclopropyl)thiophene
2-Bromo-5-(cyclopropyl)thiophene
Vzorec: C₇H₇BrS
MW: 203,099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12033270
CAS číslo: 29481-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR