Tisk…

2-(Cyclopropyl)thiophene

Dodavatel: Apollo Scientific
Varování

OR40805-1G OR40805-250MG OR40805-5G
APOSOR40805-1GEA 29130 CZK
APOSOR40805-1G APOSOR40805-250MG APOSOR40805-5G
2-(Cyclopropyl)thiophene
2-(Cyclopropyl)thiophene
Vzorec: C₇H₈S
MW: 124,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22049303
CAS číslo: 29481-22-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR