(2,4-Dichloro-3,5-dimethylphenoxy)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR40781-1G
OR40781-250MG
APOSOR40781-1GEA
3870
CZK
APOSOR40781-1G
APOSOR40781-250MG
(2,4-Dichloro-3,5-dimethylphenoxy)acetic acid
(2,4-Dichloro-3,5-dimethylphenoxy)acetic acid
Vzorec:
C₁₀H₁₀Cl₂O₃ MW: 249,091 g/mol Bod tání: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD28102587 CAS číslo: 19545-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...