Tisk…

4-Bromo-2-cyclopropylaniline 95%

Dodavatel: Apollo Scientific
Varování

OR40778-1G OR40778-250MG OR40778-5G
APOSOR40778-1GEA 14060 CZK
APOSOR40778-1G APOSOR40778-250MG APOSOR40778-5G
4-Bromo-2-cyclopropylaniline 95%
4-Bromo-2-cyclopropylaniline
Vzorec: C₉H₁₀BrN
MW: 212.086 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21099984
CAS číslo: 1353854-39-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR