(2R)-4-Iodo-2-isopropyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR40694-1G
OR40694-250MG
APOSOR40694-1GEA
21820
CZK
APOSOR40694-1G
APOSOR40694-250MG
(2R)-4-Iodo-2-isopropyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
(2R)-4-Iodo-2-isopropyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₁₀H₁₃IN₂ MW: 288,128 g/mol Storage Temperature: Ambient |
CAS číslo:
2366997-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...