Tisk…

4-Bromo-2-iodobenzaldehyde 97%

Dodavatel: Apollo Scientific
Varování

OR40677-1G OR40677-250MG OR40677-5G
APOSOR40677-1GEA 992 CZK
APOSOR40677-1G APOSOR40677-250MG APOSOR40677-5G
4-Bromo-2-iodobenzaldehyde 97%
4-Bromo-2-iodobenzaldehyde
Vzorec: C₇H₄BrIO
Storage Temperature: Ambient
MDL Number: MFCD18391619
CAS číslo: 1261470-87-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR