2-Amino-3-chlorobenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-6-formylaniline, 3-Chloroanthranilaldehyde
OR40598-250MG
OR40598-1G
APOSOR40598-250MGEA
1210
CZK
APOSOR40598-250MG
APOSOR40598-1G
2-Amino-3-chlorobenzaldehyde 98%
2-Amino-3-chlorobenzaldehyde
Vzorec:
C₇H₆ClNO MW: 155,582 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13193431 CAS číslo: 397322-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...