Tisk…

2-Bromo-6-chlorobenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Bromo-2-(bromomethyl)-3-chlorobenzene, 2-Chloro-α,6-dibromotoluene

OR40565-5G
APOSOR40565-5GEA 3810 CZK
APOSOR40565-5G
2-Bromo-6-chlorobenzyl bromide 97%
2-Bromo-6-chlorobenzyl bromide
Vzorec: C₇H₅Br₂Cl
MW: 284,376 g/mol
Bod tání: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD04038394
CAS číslo: 75002-98-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR