6-Hydroxy-3-methyl-7-nitro-1,2-benzisoxazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Methyl-7-nitro-1,2-benzisoxazol-6-ol, 6-Hydroxy-3-methyl-7-nitrobenzo[d]isoxazole
OR40444-1G
OR40444-500MG
APOSOR40444-1GEA
24600
CZK
APOSOR40444-1G
APOSOR40444-500MG
6-Hydroxy-3-methyl-7-nitro-1,2-benzisoxazole
6-Hydroxy-3-methyl-7-nitro-1,2-benzisoxazole
Vzorec:
C₈H₆N₂O₄ MW: 194,144 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14586346 CAS číslo: 112429-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...