Tisk…

7-Bromoindan-1-one

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Bromo-1-indanone, 7-Bromo-2,3-dihydro-1H-inden-1-one, 7-Bromo-2,3-dihydro-1-oxo-1H-indene

OR40392-1G OR40392-5G
APOSOR40392-1GEA 2110 CZK
APOSOR40392-1G APOSOR40392-5G
7-Bromoindan-1-one
7-Bromoindan-1-one
Vzorec: C₉H₇BrO
MW: 211,055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796574
CAS číslo: 125114-77-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR