1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(5-Methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-ol, 4-(1-Hydroxyethyl)-5-methyl-3-phenylisoxazole
OR40374-1G
APOSOR40374-1GEA
3540
CZK
APOSOR40374-1G
1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol
1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol
Vzorec:
C₁₂H₁₃NO₂ MW: 203,237 g/mol Bod tání: 101…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD16631269 CAS číslo: 6497-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...