Tisk…

Methyl 3,4-dihydro-2-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylate

Dodavatel: Apollo Scientific
Varování

OR40371-1G OR40371-250MG
APOSOR40371-1GEA 9720 CZK
APOSOR40371-1G APOSOR40371-250MG
Methyl 3,4-dihydro-2-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylate
Methyl 3,4-dihydro-2-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylate
Vzorec: C₁₁H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD16631263
CAS číslo: 1257535-36-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR