Methyl 3-chloro-6-methoxypyridine-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-chloro-6-methoxypicolinate, 3-Chloro-6-methoxy-2-(methoxycarbonyl)pyridine
OR40357-1G
OR40357-250MG
APOSOR40357-1GEA
2610
CZK
APOSOR40357-1G
APOSOR40357-250MG
Methyl 3-chloro-6-methoxypyridine-2-carboxylate
Methyl 3-chloro-6-methoxypyridine-2-carboxylate
Vzorec:
C₈H₈ClNO₃ MW: 201,607 g/mol Bod varu: 80…84 °C (0,5 mm Hg) Bod tání: 32…34 °C Storage Temperature: Ambient |
MDL Number:
MFCD14698010 CAS číslo: 175965-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...