N-(2,4-Dimethylbenzyl)cyclopropylamine hydrochloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-[(Cyclopropylamino)methyl]-m-xylene hydrochloride, N-Cyclopropyl-2,4-dimethylbenzylamine hydrochloride
OR40317-1G
OR40317-250MG
APOSOR40317-1GEA
13000
CZK
APOSOR40317-1G
APOSOR40317-250MG
N-(2,4-Dimethylbenzyl)cyclopropylamine hydrochloride
N-(2,4-Dimethylbenzyl)cyclopropylamine hydrochloride
Vzorec:
C₁₂H₁₇N·ClH MW: 211,731 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD17168267 CAS číslo: 1257535-57-1 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...