Tisk…
(2R)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-methylbutanoic acid
Dodavatel: Apollo Scientific
Synonyms: (2R)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-methylbutyric acid, (R)-2-(((tert-Butoxycarbonyl)amino)methyl)-3-methylbutanoic acid
OR40216-100MG OR40216-250MG
APOSOR40216-100MGEA 3940 CZK
APOSOR40216-100MG APOSOR40216-250MG
(2R)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-methylbutanoic acid
(2R)-2-{[(tert-Butoxycarbonyl)amino]methyl}-3-methylbutanoic acid
Vzorec: C₁₁H₂₁NO₄
MW: 231.289 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07372880
CAS číslo: 191664-14-9