Tisk…

1,3-Dimethyl-1H-indazole-6-boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (1,3-Dimethyl-1H-indazol-6-yl)boronic acid, 6-Borono-1,3-dimethyl-1H-indazole

OR40165-250MG OR40165-100MG
APOSOR40165-250MGEA 1660 CZK
APOSOR40165-250MG APOSOR40165-100MG
1,3-Dimethyl-1H-indazole-6-boronic acid
1,3-Dimethyl-1H-indazole-6-boronic acid
Vzorec: C₉H₁₁BN₂O₂
MW: 190,007 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028657
CAS číslo: 1310405-37-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR