6-Bromo-2,5-dimethyl-2H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
OR40144-1G
OR40144-250MG
APOSOR40144-1GEA
7440
CZK
APOSOR40144-1G
APOSOR40144-250MG
6-Bromo-2,5-dimethyl-2H-indazole
6-Bromo-2,5-dimethyl-2H-indazole
Vzorec:
C₉H₉BrN₂ MW: 225,085 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12028639 CAS číslo: 1159511-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...