Tisk…

6-Bromo-1,5-dimethyl-1H-indazole

Dodavatel: Apollo Scientific
Varování

OR40132-1G OR40132-250MG
APOSOR40132-1GEA 3310 CZK
APOSOR40132-1G APOSOR40132-250MG
6-Bromo-1,5-dimethyl-1H-indazole
6-Bromo-1,5-dimethyl-1H-indazole
Vzorec: C₉H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12028628
CAS číslo: 1159511-83-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR