4-Hydroxy-3-(methoxycarbonyl)benzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Methyl 4-hydroxyisophthalate, Methyl 5-carboxy-2-hydroxybenzoate
OR40106-1G
OR40106-250MG
APOSOR40106-1GEA
16140
CZK
APOSOR40106-1G
APOSOR40106-250MG
4-Hydroxy-3-(methoxycarbonyl)benzoic acid 95%
4-Hydroxy-3-(methoxycarbonyl)benzoic acid
Vzorec:
C₉H₈O₅ MW: 196,157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11869747 CAS číslo: 41684-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...