Tisk…

3-Iodo-5-nitrobenzonitrile 95%

Dodavatel: Apollo Scientific
Varování

OR401059-1G OR401059-250MG OR401059-5G
APOSOR401059-1GEA 14880 CZK
APOSOR401059-1G APOSOR401059-250MG APOSOR401059-5G
3-Iodo-5-nitrobenzonitrile 95%
3-Iodo-5-nitrobenzonitrile
Vzorec: C₇H₃IN₂O₂
MW: 274.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464943
CAS číslo: 49674-30-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR