Tisk…

2-Iodo-3-nitrobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR401054-1G OR401054-250MG OR401054-5G
APOSOR401054-1GEA 1660 CZK
APOSOR401054-1G APOSOR401054-250MG APOSOR401054-5G
2-Iodo-3-nitrobenzyl bromide 95%
2-Iodo-3-nitrobenzyl bromide
Vzorec: C₇H₅BrINO₂
MW: 341,929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398002
CAS číslo: 1261777-67-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR