Tisk…

2-Iodo-3-nitrobenzonitrile 95%

Dodavatel: Apollo Scientific
Varování

OR401051-1G OR401051-250MG OR401051-5G
APOSOR401051-1GEA 1660 CZK
APOSOR401051-1G APOSOR401051-250MG APOSOR401051-5G
2-Iodo-3-nitrobenzonitrile 95%
2-Iodo-3-nitrobenzonitrile
Vzorec: C₇H₃IN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18397968
CAS číslo: 1261481-69-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR