Tisk…

3-Chloro-5-nitrobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-3-chloro-5-nitrobenzene

OR401031-1G OR401031-250MG
APOSOR401031-1GEA 20270 CZK
APOSOR401031-1G APOSOR401031-250MG
3-Chloro-5-nitrobenzyl bromide
3-Chloro-5-nitrobenzyl bromide
Vzorec: C₇H₅BrClNO₂
MW: 250,477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505896
CAS číslo: 865624-17-5
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR