Tisk…

3-Chloro-2-nitrobenzaldehyde

Dodavatel: Apollo Scientific

OR401017-1G
APOSOR401017-1GEA 4070 CZK
APOSOR401017-1G
3-Chloro-2-nitrobenzaldehyde
3-Chloro-2-nitrobenzaldehyde
Vzorec: C₇H₄ClNO₃
MW: 185,565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797953
CAS číslo: 22233-52-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR