Tisk…

3-Chloro-4-iodobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR400981-1G OR400981-250MG OR400981-5G
APOSOR400981-1GEA 1660 CZK
APOSOR400981-1G APOSOR400981-250MG APOSOR400981-5G
3-Chloro-4-iodobenzyl bromide 95%
3-Chloro-4-iodobenzyl bromide
Vzorec: C₇H₅BrClI
Storage Temperature: Ambient
MDL Number: MFCD18393368
CAS číslo: 166386-59-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR