Tisk…

4-Chloro-3-iodobenzaldehyde

Dodavatel: Apollo Scientific

OR400973-250MG OR400973-1G
APOSOR400973-250MGEA 2030 CZK
APOSOR400973-250MG APOSOR400973-1G
4-Chloro-3-iodobenzaldehyde
4-Chloro-3-iodobenzaldehyde
Vzorec: C₇H₄ClIO
MW: 266,464 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836681
CAS číslo: 276866-90-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR