3-Chloro-4-iodobenzoic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR400960-1G
OR400960-5G
OR400960-25G
APOSOR400960-1GEA
622
CZK
APOSOR400960-1G
APOSOR400960-5G
APOSOR400960-25G
3-Chloro-4-iodobenzoic acid 96%
3-Chloro-4-iodobenzoic acid
Vzorec:
C₇H₄ClIO₂ MW: 282,463 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06637386 CAS číslo: 58123-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...