Methyl 2-bromo-4-iodobenzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR400949-1G
OR400949-5G
OR400949-25G
APOSOR400949-1GEA
622
CZK
APOSOR400949-1G
APOSOR400949-5G
APOSOR400949-25G
Methyl 2-bromo-4-iodobenzoate 95%
Methyl 2-bromo-4-iodobenzoate
Vzorec:
C₈H₆BrIO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11870175 CAS číslo: 1261588-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...