Tisk…

Benzo[b]thiophene-4-carboxylic acid

Dodavatel: Apollo Scientific

OR400932-1G
APOSOR400932-1GEA 3740 CZK
APOSOR400932-1G
Benzo[b]thiophene-4-carboxylic acid
Benzo[b]thiophene-4-carboxylic acid
Vzorec: C₉H₆O₂S
MW: 178,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01929340
CAS číslo: 10134-95-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR