Tisk…

2-Amino-6-bromo-3-iodophenol

Dodavatel: Apollo Scientific
Varování

OR400890-1G OR400890-250MG OR400890-5G
APOSOR400890-1GEA 42980 CZK
APOSOR400890-1G APOSOR400890-250MG APOSOR400890-5G
2-Amino-6-bromo-3-iodophenol
2-Amino-6-bromo-3-iodophenol
Vzorec: C₆H₅BrINO
Storage Temperature: Ambient
MDL Number: MFCD30730278
CAS číslo: 2092616-33-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR