Tisk…

6-Bromo-3-iodo-2-nitrophenol

Dodavatel: Apollo Scientific
Varování

OR400888-250MG OR400888-1G
APOSOR400888-250MGEA 4960 CZK
APOSOR400888-250MG APOSOR400888-1G
6-Bromo-3-iodo-2-nitrophenol
6-Bromo-3-iodo-2-nitrophenol
Vzorec: C₆H₃BrINO₃
MW: 343,901 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30730276
CAS číslo: 2090513-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR