Tisk…

4-Bromo-5-iodobenzene-1,2-diamine 95%

Dodavatel: Apollo Scientific
Varování

OR400871-1G OR400871-250MG
APOSOR400871-1GEA 20470 CZK
APOSOR400871-1G APOSOR400871-250MG
4-Bromo-5-iodobenzene-1,2-diamine 95%
4-Bromo-5-iodobenzene-1,2-diamine
Vzorec: C₆H₆BrIN₂
Storage Temperature: Ambient
MDL Number: MFCD30730274
CAS číslo: 2149591-36-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR