Tisk…

2-Bromo-3-chloro-6-hydroxybenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

OR400793-1G OR400793-250MG
APOSOR400793-1GEA 18620 CZK
APOSOR400793-1G APOSOR400793-250MG
2-Bromo-3-chloro-6-hydroxybenzoic acid 95%
2-Bromo-3-chloro-6-hydroxybenzoic acid
Vzorec: C₇H₄BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD29037278
CAS číslo: 1934463-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR