Tisk…

2-Bromo-5-formylbenzoic acid

Dodavatel: Apollo Scientific
Varování

OR400784-1G OR400784-250MG OR400784-5G
APOSOR400784-1GEA 4140 CZK
APOSOR400784-1G APOSOR400784-250MG APOSOR400784-5G
2-Bromo-5-formylbenzoic acid
2-Bromo-5-formylbenzoic acid
Vzorec: C₈H₅BrO₃
MW: 229,027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26855793
CAS číslo: 1289007-84-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR