6-Amino-2-bromo-3-chlorobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR400780-1G
OR400780-250MG
APOSOR400780-1GEA
20470
CZK
APOSOR400780-1G
APOSOR400780-250MG
6-Amino-2-bromo-3-chlorobenzonitrile
6-Amino-2-bromo-3-chlorobenzonitrile
Vzorec:
C₇H₄BrClN₂ MW: 231.477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29037275 CAS číslo: 1934421-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...