Tisk…

4-Bromo-3-formylbenzoic acid

Dodavatel: Apollo Scientific
Varování

OR400762-1G OR400762-250MG
APOSOR400762-1GEA 12400 CZK
APOSOR400762-1G APOSOR400762-250MG
4-Bromo-3-formylbenzoic acid
4-Bromo-3-formylbenzoic acid
Vzorec: C₈H₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD20485838
CAS číslo: 776315-23-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR