Tisk…

2-Bromo-4-iodonitrobenzene

Dodavatel: Apollo Scientific
Varování

OR400757-1G OR400757-250MG
APOSOR400757-1GEA 23150 CZK
APOSOR400757-1G APOSOR400757-250MG
2-Bromo-4-iodonitrobenzene
2-Bromo-4-iodonitrobenzene
Vzorec: C₆H₃BrINO₂
MW: 327,902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391757
CAS číslo: 860603-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR