Tisk…

Methyl 2-bromo-3-formylbenzoate

Dodavatel: Apollo Scientific
Varování

OR400756-1G OR400756-250MG OR400756-5G
APOSOR400756-1GEA 16530 CZK
APOSOR400756-1G APOSOR400756-250MG APOSOR400756-5G
Methyl 2-bromo-3-formylbenzoate
Methyl 2-bromo-3-formylbenzoate
Vzorec: C₉H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD11110489
CAS číslo: 750585-94-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR