Tisk…

3-Bromo-2-iodobenzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

OR400746-1G OR400746-250MG OR400746-5G
APOSOR400746-1GEA 16340 CZK
APOSOR400746-1G APOSOR400746-250MG APOSOR400746-5G
3-Bromo-2-iodobenzaldehyde 95%
3-Bromo-2-iodobenzaldehyde
Vzorec: C₇H₄BrIO
MW: 310,915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391618
CAS číslo: 1261860-40-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR