2-Bromo-3-iodobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR400744-1G
OR400744-250MG
APOSOR400744-1GEA
16340
CZK
APOSOR400744-1G
APOSOR400744-250MG
2-Bromo-3-iodobenzonitrile
2-Bromo-3-iodobenzonitrile
Vzorec:
C₇H₃BrIN MW: 307,914 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18391642 CAS číslo: 1261498-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...