Tisk…

5-Bromo-2-chlorophenacyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR400708-250MG OR400708-1G OR400708-5G
APOSOR400708-250MGEA 6020 CZK
APOSOR400708-250MG APOSOR400708-1G APOSOR400708-5G
5-Bromo-2-chlorophenacyl bromide 97%
5-Bromo-2-chlorophenacyl bromide
Vzorec: C₈H₅Br₂ClO
MW: 312,386 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392520
CAS číslo: 1261775-99-8
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR