Tisk…

2-Bromo-5-nitrobenzenesulphonyl chloride

Dodavatel: Apollo Scientific

OR400707-1G OR400707-250MG OR400707-5G
APOSOR400707-1GEA 13360 CZK
APOSOR400707-1G APOSOR400707-250MG APOSOR400707-5G
2-Bromo-5-nitrobenzenesulphonyl chloride
2-Bromo-5-nitrobenzenesulphonyl chloride
Vzorec: C₆H₃BrClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD06254509
CAS číslo: 98130-55-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR