Tisk…

4-Amino-2-bromobenzoic acid

Dodavatel: Apollo Scientific

OR400700-1G OR400700-250MG OR400700-5G
APOSOR400700-1GEA 7250 CZK
APOSOR400700-1G APOSOR400700-250MG APOSOR400700-5G
4-Amino-2-bromobenzoic acid
4-Amino-2-bromobenzoic acid
Vzorec: C₇H₆BrNO₂
MW: 216,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01569396
CAS číslo: 2486-52-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR