5-Bromo-2-nitrobenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400698-1G
OR400698-5G
APOSOR400698-1GEA
8240
CZK
APOSOR400698-1G
APOSOR400698-5G
5-Bromo-2-nitrobenzenesulphonyl chloride
5-Bromo-2-nitrobenzenesulphonyl chloride
Vzorec:
C₆H₃BrClNO₄S Bod tání: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD12912807 CAS číslo: 1193388-84-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...