Tisk…

4-Bromo-3-methoxyphenylene-1,2-diamine

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-3-methoxybenzene-1,2-diamine

OR400692-1G OR400692-250MG
APOSOR400692-1GEA 20470 CZK
APOSOR400692-1G APOSOR400692-250MG
4-Bromo-3-methoxyphenylene-1,2-diamine
4-Bromo-3-methoxyphenylene-1,2-diamine
Vzorec: C₇H₉BrN₂O
MW: 217,063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24643738
CAS číslo: 1162696-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR