4-Bromo-2-iodo-6-nitroanisole
Dodavatel: Apollo Scientific
|
Varování
|
OR400691-1G
OR400691-5G
APOSOR400691-1GEA
5200
CZK
APOSOR400691-1G
APOSOR400691-5G
4-Bromo-2-iodo-6-nitroanisole
4-Bromo-2-iodo-6-nitroanisole
Vzorec:
C₇H₅BrINO₃ MW: 357,928 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28898061 CAS číslo: 1935459-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...